Merck
  • Differential Effects of Tacrolimus versus Sirolimus on the Proliferation, Activation and Differentiation of Human B Cells.
Differential Effects of Tacrolimus versus Sirolimus on the Proliferation, Activation and Differentiation of Human B Cells.
PloS one (2015-06-19)
Opas Traitanon, James M Mathew, Giovanna La Monica, Luting Xu, Valeria Mas, Lorenzo Gallon
ZUSAMMENFASSUNG
The direct effect of immunosuppressive drugs calcineurin inhibitor (Tacrolimus, TAC) and mTOR inhibitor (Sirolimus, SRL) on B cell activation, differentiation and proliferation is not well documented. Purified human B cells from healthy volunteers were stimulated through the B Cell Receptor with Anti-IgM + anti-CD40 + IL21 in the absence / presence of TAC or SRL. A variety of parameters of B cell activity including activation, differentiation, cytokine productions and proliferation were monitored by flow cytometry. SRL at clinically relevant concentrations (6 ng/ml) profoundly inhibited CD19(+ )B cell proliferation compared to controls whereas TAC at similar concentrations had a minimal effect. CD27(+) memory B cells were affected more by SRL than naïve CD27- B cells. SRL effectively blocked B cell differentiation into plasma cells (CD19(+)CD138(+) and Blimp1(+)/Pax5(low) cells) even at low dose (2 ng/ml), and totally eliminated them at 6 ng/ml. SRL decreased absolute B cell counts, but the residual responding cells acquired an activated phenotype (CD25(+)/CD69(+)) and increased the expression of HLA-DR. SRL-treated stimulated B cells on a per cell basis were able to enhance the proliferation of allogeneic CD4(+)CD25(-) T cells and induce a shift toward the Th1 phenotype. Thus, SRL and TAC have different effects on B lymphocytes. These data may provide insights into the clinical use of these two agents in recipients of solid organ transplants.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Rapamycin aus Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
5(6)-Carboxyfluoreszeindiacetat-N-succinimidylester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
FK-506 Monohydrat, ≥98% (HPLC)
Sigma-Aldrich
5-Carboxy-Fluoresceindiacetat N-Succinimidylester, suitable for fluorescence, ≥95.0% (HPLC)