Merck
Cultivation of Chlorella sp. GD using piggery wastewater for biomass and lipid production.
Bioresource technology (2015-07-27)
Chiu-Mei Kuo, Tsai-Yu Chen, Tsung-Hsien Lin, Chien-Ya Kao, Jinn-Tsyy Lai, Jo-Shu Chang, Chih-Sheng Lin
ZUSAMMENFASSUNG
The development of a culture system for Chlorella sp. GD to efficiently produce biomass and oil for biodiesel production was investigated. Chlorella sp. GD was cultivated with 0%, 25%, 50%, 75% and 100% piggery wastewater (diluted by medium) at 300 μmol m(-2) s(-1), a 2% CO2 aeration rate of 0.2 vvm and 26±1°C; after a 10-day culture in batch cultures, the maximum specific growth rate and biomass productivity of the microalga obtained in 100% piggery wastewater were 0.839 d(-1) and 0.681 g L(-1) d(-1), respectively. The highest lipid content and lipid productivity were 29.3% and 0.155 g L(-1) d(-1) at 25% wastewater, respectively. In semi-continuous cultures, the biomass and lipid productivities with 25-75% wastewater ratios were greater than 0.852 and 0.128 g L(-1) d(-1), respectively. These results show that Chlorella sp. GD grows efficiently in piggery wastewater, and that a stable growth performance was achieved for long-term microalgal cultivation in a semi-continuous culture.