Merck
  • The covalently bonded cellulose tris(3,5-dimethylphenylcarbamate) on a silica monolithic capillary column for enantioseparation in capillary electrochromatography.
The covalently bonded cellulose tris(3,5-dimethylphenylcarbamate) on a silica monolithic capillary column for enantioseparation in capillary electrochromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-09-02)
Xiaoli Dong, Ren'an Wu, Jing Dong, Minghuo Wu, Yan Zhu, Hanfa Zou
ZUSAMMENFASSUNG
A chiral capillary monolithic column for capillary electrochromatography (CEC) was prepared by covalent bonding of cellulose tris(3,5-dimethylphenylcarbamate) (CDMPC) on the silica monolithic matrix within the confine of a 50-microm i.d. bare fused silica capillary. Several pairs of enantiomers including neutral and basic analytes were baseline resolved on the newly prepared chiral capillary monolithic column in CEC with aqueous mobile phases. Fast enantioseparation was achieved due to the favorable dynamic properties of silica monolith. The covalent bonding of CDMPC as the chiral stationary phase for CEC also enabled the use of THF in mobile phase for enantioseparation of prazquantel by overcoming the incompatibility of THF and the physically coated CDMPC on a column.