Merck
  • Methylation-associated silencing of heparan sulfate D-glucosaminyl 3-O-sulfotransferase-2 (3-OST-2) in human breast, colon, lung and pancreatic cancers.
Methylation-associated silencing of heparan sulfate D-glucosaminyl 3-O-sulfotransferase-2 (3-OST-2) in human breast, colon, lung and pancreatic cancers.
Oncogene (2003-01-16)
Kazuaki Miyamoto, Kiyoshi Asada, Takashi Fukutomi, Eriko Okochi, Yukiko Yagi, Tadashi Hasegawa, Toshimasa Asahara, Takashi Sugimura, Toshikazu Ushijima
ZUSAMMENFASSUNG
Aberrant CpG methylations play important roles in cancer development and progression. In this study, aberrant methylations in human breast cancer were searched for using methylation-sensitive representational difference analysis (MS-RDA). A CpG island (CGI) in the 5' region of the heparan sulfate D-glucosaminyl 3-O-sulfotransferase-2 (3-OST-2) gene was found to be hypermethylated, while its exon 2 was hypomethylated. In seven breast cancer cell lines, hypermethylation of the 5' region and loss of 3-OST-2 expression were observed. Treatment with a demethylating agent, 5-aza-2'-deoxycytidine, removed the methylation of the CGI in the 5' region and restored its expression, demonstrating silencing of the 3-OST-2 gene. Methylation-specific PCR (MSP) analysis in 85 primary breast cancers showed that the hypermethylation of the CGI in the 5' region was present in 75 (88%) of them. Quantitative reverse transcriptase-PCR (RT-PCR) analysis in 37 primary breast cancers showed that the average expression level was decreased in them. Further, MSP analysis in primary colon, lung and pancreatic cancers showed that hypermethylation of the CGI in the 5' region was present in the colon (8/10, 80%), lung (7/10, 70%) and pancreatic (10/10, 100%) cancers. These results showed that silencing of 3-OST-2 was present in a wide range of human cancers. The 3-OST-2 gene encodes an enzyme involved in the final modification step of heparan sulfate proteoglycans (HSPGs), and its silencing is expected to result in abnormal modification of HSPGs and abnormal signal transduction. From the high incidence, silencing of the 3-OST-2 gene is expected to have high diagnostic, and potentially therapeutic, values.