Merck
  • Iodine monochloride facilitated deglycosylation, anomerization, and isomerization of 3-substituted thymidine analogues.
Iodine monochloride facilitated deglycosylation, anomerization, and isomerization of 3-substituted thymidine analogues.
Nucleosides, nucleotides & nucleic acids (2014-11-06)
Ahmed Khalil, Keisuke Ishita, Tehane Ali, Rohit Tiwari, Ramy Riachy, Antonio Toppino, Sherifa Hasabelnaby, Naum Sayfullin, Allen G Oliver, Judith Gallucci, Zhenguo Huang, Werner Tjarks
ZUSAMMENFASSUNG
The reaction of thymidine, 3-mono-, and 3,3',5'-trialkylsubstitued thymidine analogues with iodine monochloride (ICl) was investigated. Treatment with ICl resulted in rapid deglycosylation, anomerization, and isomerization of thymidine and 3-substituted thymidine analogues under various reaction conditions leading to the formation of the nucleobases as the major products accompanied by minor formation of α-furanosidic-, α-pyranosidic-, and β-pyranosidic nucleosides. On the other hand, 3,3',5'-trisubstitued thymidine analogues were only deglycosylated and anomerized. These results are similar to those observed for the acidic hydrolysis of the glycoside bond in nucleosides, but were presumably caused by the Lewis acid character of an iodine electrophile.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloriod, reagent grade, ≥95%
Sigma-Aldrich
Chloriod, ACS reagent, 1.10±0.1 I/Cl ratio basis
Sigma-Aldrich
Iodmonochlorid -Lösung, 1.0 M in methylene chloride
Sigma-Aldrich
Chloriod, 99.998% trace metals basis