Merck
  • Two Carbonylations of Methyl Iodide and Trimethylamine to Acetic acid and N, N-Dimethylacetamide by Rhodium (I) Complex: Stability of Rhodium (I) Complex under Anhydrous Condition.
Two Carbonylations of Methyl Iodide and Trimethylamine to Acetic acid and N, N-Dimethylacetamide by Rhodium (I) Complex: Stability of Rhodium (I) Complex under Anhydrous Condition.
Catalysts (2015)
Hong JH.