Merck
  • New and highly effective method for the oxidation of primary and secondary alcohols to carbonyl compounds.
New and highly effective method for the oxidation of primary and secondary alcohols to carbonyl compounds.
Journal of the American Chemical Society (1972)
Corey EJ and Kim CU.