Merck
  • Copper (I) Oxide-Mediated Cyclization of o-Haloaryl N-Tosylhydrazones: Efficient Synthesis of Indazoles.
Copper (I) Oxide-Mediated Cyclization of <I>o</I>-Haloaryl<I> N</I>-Tosylhydrazones: Efficient Synthesis of Indazoles.
Advanced Synthesis &amp; Catalysis (2016)
Tang M, et al.