Merck
  • Characterization of an unusual reaction product of tin (II) chloride with sodium N, N-diethyldithiocarbamate.
Characterization of an unusual reaction product of tin (II) chloride with sodium <I>N, N</I>-diethyldithiocarbamate.
Polyhedron (1994)
Selvaraju R, et al.