Merck
  • Concentration of Sec12 at ER exit sites via interaction with cTAGE5 is required for collagen export.
Concentration of Sec12 at ER exit sites via interaction with cTAGE5 is required for collagen export.
The Journal of cell biology (2014-09-10)
Kota Saito, Koh Yamashiro, Noriko Shimazu, Tomoya Tanabe, Kenji Kontani, Toshiaki Katada
ZUSAMMENFASSUNG
Mechanisms for exporting variably sized cargo from the endoplasmic reticulum (ER) using the same machinery remain poorly understood. COPII-coated vesicles, which transport secretory proteins from the ER to the Golgi apparatus, are typically 60-90 nm in diameter. However, collagen, which forms a trimeric structure that is too large to be accommodated by conventional transport vesicles, is also known to be secreted via a COPII-dependent process. In this paper, we show that Sec12, a guanine-nucleotide exchange factor for Sar1 guanosine triphosphatase, is concentrated at ER exit sites and that this concentration of Sec12 is specifically required for the secretion of collagen VII but not other proteins. Furthermore, Sec12 recruitment to ER exit sites is organized by its direct interaction with cTAGE5, a previously characterized collagen cargo receptor component, which functions together with TANGO1 at ER exit sites. These findings suggest that the export of large cargo requires high levels of guanosine triphosphate-bound Sar1 generated by Sec12 localized at ER exit sites.