Merck
  • Sphingosylphosphorylcholine and lysophosphatidylcholine are ligands for the G protein-coupled receptor GPR4.
Sphingosylphosphorylcholine and lysophosphatidylcholine are ligands for the G protein-coupled receptor GPR4.
The Journal of biological chemistry (2006-02-25)
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Avanti
Lyso SM (d18:1), Avanti Research - A Croda Brand