Merck
  • A hypermorphic IkappaBalpha mutation is associated with autosomal dominant anhidrotic ectodermal dysplasia and T cell immunodeficiency.
A hypermorphic IkappaBalpha mutation is associated with autosomal dominant anhidrotic ectodermal dysplasia and T cell immunodeficiency.
The Journal of clinical investigation (2003-10-03)
Gilles Courtois, Asma Smahi, Janine Reichenbach, Rainer Döffinger, Caterina Cancrini, Marion Bonnet, Anne Puel, Christine Chable-Bessia, Shoji Yamaoka, Jacqueline Feinberg, Sophie Dupuis-Girod, Christine Bodemer, Susanna Livadiotti, Francesco Novelli, Paolo Rossi, Alain Fischer, Alain Israël, Arnold Munnich, Françoise Le Deist, Jean-Laurent Casanova
ZUSAMMENFASSUNG
X-linked anhidrotic ectodermal dysplasia with immunodeficiency (XL-EDA-ID) is caused by hypomorphic mutations in the gene encoding NEMO/IKKgamma, the regulatory subunit of the IkappaB kinase (IKK) complex. IKK normally phosphorylates the IkappaB-inhibitors of NF-kappaB at specific serine residues, thereby promoting their ubiquitination and degradation by the proteasome. This allows NF-kappaB complexes to translocate into the nucleus where they activate their target genes. Here, we describe an autosomal-dominant (AD) form of EDA-ID associated with a heterozygous missense mutation at serine 32 of IkappaBalpha. This mutation is gain-of-function, as it enhances the inhibitory capacity of IkappaBalpha by preventing its phosphorylation and degradation, and results in impaired NF-kappaB activation. The developmental, immunologic, and infectious phenotypes associated with hypomorphic NEMO and hypermorphic IKBA mutations largely overlap and include EDA, impaired cellular responses to ligands of TIR (TLR-ligands, IL-1beta, and IL-18), and TNFR (TNF-alpha, LTalpha1/beta2, and CD154) superfamily members and severe bacterial diseases. However, AD-EDA-ID but not XL-EDA-ID is associated with a severe and unique T cell immunodeficiency. Despite a marked blood lymphocytosis, there are no detectable memory T cells in vivo, and naive T cells do not respond to CD3-TCR activation in vitro. Our report highlights both the diversity of genotypes associated with EDA-ID and the diversity of immunologic phenotypes associated with mutations in different components of the NF-kappaB signaling pathway.