Merck
  • Disruption of Nrf2/ARE signaling impairs antioxidant mechanisms and promotes cell degradation pathways in aged skeletal muscle.
Disruption of Nrf2/ARE signaling impairs antioxidant mechanisms and promotes cell degradation pathways in aged skeletal muscle.
Biochimica et biophysica acta (2012-03-01)
Corey J Miller, Sellamuthu S Gounder, Sankaranarayanan Kannan, Karan Goutam, Vasanthi R Muthusamy, Matthew A Firpo, J David Symons, Robert Paine, John R Hoidal, Namakkal Soorappan Rajasekaran
ZUSAMMENFASSUNG
Age-associated decline in antioxidant potential and accumulation of reactive oxygen/nitrogen species are primary causes for multiple health problems, including muscular dystrophy and sarcopenia. The role of the nuclear erythroid-2-p45-related factor-2 (Nrf2) signaling has been implicated in antioxidant gene regulation. Here, we investigated the loss-of-function mechanisms for age-dependent regulation of Nrf2/ARE (Antioxidant Response Element) signaling in skeletal muscle (SM). Under basal physiological conditions, disruption of Nrf2 showed minimal effects on antioxidant defenses in young (2months) Nrf2-/- mice. Interestingly, mRNA and protein levels of NADH Quinone Oxidase-1 were dramatically (*P<0.001) decreased in Nrf2-/- SM when compared to WT at 2months of age, suggesting central regulation of NQO1 occurs through Nrf2. Subsequent analysis of the Nrf2-dependent transcription and translation showed that the aged mice (>24months) had a significant increase in ROS along with a decrease in glutathione (GSH) levels and impaired antioxidants in Nrf2-/- when compared to WT SM. Further, disruption of Nrf2 appears to induce oxidative stress (increased ROS, HNE-positive proteins), ubiquitination and pro-apoptotic signals in the aged SM of Nrf2-/- mice. These results indicate a direct role for Nrf2/ARE signaling on impairment of antioxidants, which contribute to muscle degradation pathways upon aging. Our findings conclude that though the loss of Nrf2 is not amenable at younger age; it could severely affect the SM defenses upon aging. Thus, Nrf2 signaling might be a potential therapeutic target to protect the SM from age-dependent accumulation of ROS by rescuing redox homeostasis to prevent age-related muscle disorders such as sarcopenia and myopathy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
meta-Phosphorsäure, ACS reagent, chips, 33.5-36.5%