Merck
  • Proapoptotic BAX and BAK modulate the unfolded protein response by a direct interaction with IRE1alpha.
Proapoptotic BAX and BAK modulate the unfolded protein response by a direct interaction with IRE1alpha.
Science (New York, N.Y.) (2006-04-29)
Claudio Hetz, Paula Bernasconi, Jill Fisher, Ann-Hwee Lee, Michael C Bassik, Bruno Antonsson, Gabriel S Brandt, Neal N Iwakoshi, Anna Schinzel, Laurie H Glimcher, Stanley J Korsmeyer
ZUSAMMENFASSUNG
Accumulation of misfolded protein in the endoplasmic reticulum (ER) triggers an adaptive stress response-termed the unfolded protein response (UPR)-mediated by the ER transmembrane protein kinase and endoribonuclease inositol-requiring enzyme-1alpha (IRE1alpha). We investigated UPR signaling events in mice in the absence of the proapoptotic BCL-2 family members BAX and BAK [double knockout (DKO)]. DKO mice responded abnormally to tunicamycin-induced ER stress in the liver, with extensive tissue damage and decreased expression of the IRE1 substrate X-box-binding protein 1 and its target genes. ER-stressed DKO cells showed deficient IRE1alpha signaling. BAX and BAK formed a protein complex with the cytosolic domain of IRE1alpha that was essential for IRE1alpha activation. Thus, BAX and BAK function at the ER membrane to activate IRE1alpha signaling and to provide a physical link between members of the core apoptotic pathway and the UPR.