Merck
  • Efficient and bright colloidal quantum dot light-emitting diodes via controlling the shell thickness of quantum dots.
Efficient and bright colloidal quantum dot light-emitting diodes via controlling the shell thickness of quantum dots.
ACS applied materials & interfaces (2013-11-07)
Huaibin Shen, Qinli Lin, Hongzhe Wang, Lei Qian, Yixing Yang, Alexandre Titov, Jake Hyvonen, Ying Zheng, Lin Song Li
ZUSAMMENFASSUNG
In this paper, we use a simple device architecture based on solution-processed ZnO nanoparticles (NPs) as the electron injection/transport layer and bilayer structure of poly(ethylenedioxythiophene):polystyrene sulfonate (PEDOT:PSS)/poly[9,9-dioctylfluorene-co-N-[4-(3-methylpropyl)]-diphenylamine] (TFB) as the hole injection/transport layer to assess the effect of shell thickness on the properties of quantum-dot-based light emitting diodes (QD-LEDs), comprising CdSe/CdS/ZnS core-shell QDs as the emitting layer. QDs with varying shell thickness were assessed to determine the best option of shell thickness, and the best improvement in device performance was observed when the shell thickness was 2.1 nm. Thereafter, different emissions of QDs, but with optimized same shell thickness (∼2.1 nm), were selected as emitters to be fabricated into same structured QD-LEDs. Highly bright orange-red and green QD-LEDs with peak luminances up to ∼30 000 and ∼52 000 cd m(-2), and power efficiencies of 16 and 19.7 lm W(-1), respectively, were demonstrated successfully. These results may demonstrate a striking basic prototype for the commercialization of QD-based displays and solid-state lightings.