Merck
  • Inhibition of Csn3 expression induces growth arrest and apoptosis of hepatocellular carcinoma cells.
Inhibition of Csn3 expression induces growth arrest and apoptosis of hepatocellular carcinoma cells.
Cancer chemotherapy and pharmacology (2012-01-13)
Yong-sheng Yu, Zheng-hao Tang, Qing-chun Pan, Xiao-hua Chen, Xue-ni Liu, Guo-qing Zang
ZUSAMMENFASSUNG
Csn3 (or CSN3) encodes the third subunit of an eight-subunit complex, the COP9 signalosome (CSN), which acts as a protein kinase and a deneddylase in mammalian cells. Previous studies have shown that Csn3 is essential for maintenance of cell proliferation in the mouse embryonic epiblast and associated with the tumorigenesis process in osteosarcoma. However, its correlation with hepatocellular carcinoma (HCC) has not been explored yet. The expression of Csn3 in HCC (n = 30), cirrhosis (n = 30), and normal tissues (n = 30) was detected using immunohistochemical analysis. The impacts of lentivirus-mediated inhibition of Csn3 on HCC cells were detected using MTT, BrdU incorporation assay, and flow cytometric analysis. In addition, the colony formation and tumor growth ability in nude mice were detected to define the role of Csn3 in tumorigenesis. Knockdown of Csn3 expression in HCC cell lines (SMMC-7721 and Hep3B) significantly inhibits the tumor growth both in vitro and in vivo. Further investigation indicates that this growth inhibition effect may be mediated through cell cycle arrest in G0/G1 phase and inductions of pro-apoptotic proteins BIK and Caspase-8. In addition, knockdown of Csn3 expression evidently suppresses tumor growth in a xenograft nude mice model. Collectively, this study demonstrates Csn3 as an oncogene that regulates the tumorigenesis process in HCC cells.