Merck
  • Ruthenium-Catalyzed Alkoxycarbonylation of Alkenes with Paraformaldehyde as a Carbon Monoxide Substitute.
Ruthenium-Catalyzed Alkoxycarbonylation of Alkenes with Paraformaldehyde as a Carbon Monoxide Substitute.
ChemCatChem (2014)
Liu Q, et al.