Merck
  • Methyl formate as a carbonylating agent for the catalytic conversion of phenol to methyl phenyl carbonate.
Methyl formate as a carbonylating agent for the catalytic conversion of phenol to methyl phenyl carbonate.
Green Chemistry (2015)
Wu C, et al.