Merck
  • An environmentally benign synthesis of isoxazolines and isoxazoles mediated by potassium chloride in water.
An environmentally benign synthesis of isoxazolines and isoxazoles mediated by potassium chloride in water.
Tetrahedron Letters (2014)
Han L, et al.