Merck
  • Effects of additional multiwall carbon nanotubes on impact behaviors of LiNi0.5Mn0.3Co0.2O2 battery electrodes.
Effects of additional multiwall carbon nanotubes on impact behaviors of LiNi<SUB>0.5</SUB>Mn<SUB>0.3</SUB>Co<SUB>0.2</SUB>O<SUB>2</SUB> battery electrodes.
Journal of Applied Physics (2015)
Le AV, et al.