Merck
  • Cholinergic dysfunction alters synaptic integration between thalamostriatal and corticostriatal inputs in DYT1 dystonia.
Cholinergic dysfunction alters synaptic integration between thalamostriatal and corticostriatal inputs in DYT1 dystonia.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2012-08-31)
Giuseppe Sciamanna, Annalisa Tassone, Georgia Mandolesi, Francesca Puglisi, Giulia Ponterio, Giuseppina Martella, Graziella Madeo, Giorgio Bernardi, David G Standaert, Paola Bonsi, Antonio Pisani
ZUSAMMENFASSUNG
Projections from thalamic intralaminar nuclei convey sensory signals to striatal cholinergic interneurons. These neurons respond with a pause in their pacemaking activity, enabling synaptic integration with cortical inputs to medium spiny neurons (MSNs), thus playing a crucial role in motor function. In mice with the DYT1 dystonia mutation, stimulation of thalamostriatal axons, mimicking a response to salient events, evoked a shortened pause and triggered an abnormal spiking activity in interneurons. This altered pattern caused a significant rearrangement of the temporal sequence of synaptic activity mediated by M(1) and M(2) muscarinic receptors in MSNs, consisting of an increase in postsynaptic currents and a decrease of presynaptic inhibition, respectively. Consistent with a major role of acetylcholine, either lowering cholinergic tone or antagonizing postsynaptic M(1) muscarinic receptors normalized synaptic activity. Our data demonstrate an abnormal time window for synaptic integration between thalamostriatal and corticostriatal inputs, which might alter the action selection process, thereby predisposing DYT1 gene mutation carriers to develop dystonic movements.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gewebe-PCR-Kit Extract-N-Amp, sufficient for 100 extractions, sufficient for 100 amplifications
Sigma-Aldrich
Gewebe-PCR-Kit Extract-N-Amp, sufficient for 1000 extractions, sufficient for 1000 amplifications