Merck
  • Assignment of the gene encoding human Krüppel-related zinc finger protein 4 (GLI4) to 8q24.3 by fluorescent in situ hybridization.
Assignment of the gene encoding human Krüppel-related zinc finger protein 4 (GLI4) to 8q24.3 by fluorescent in situ hybridization.
Cytogenetics and cell genetics (1996-01-01)
K Kas, I Wlodarska, E Meyen, H Van den Berghe, W J Van de Ven
ZUSAMMENFASSUNG
The gene for human Krüppel-related protein 4, (HKR4, gene symbol GLI4), a zinc finger protein of unknown function, has been localized by fluorescence in situ hybridization to chromosome 8q24.3, distal to c-MYC.