Merck
  • Arhgef16, a novel Elmo1 binding partner, promotes clearance of apoptotic cells via RhoG-dependent Rac1 activation.
Arhgef16, a novel Elmo1 binding partner, promotes clearance of apoptotic cells via RhoG-dependent Rac1 activation.
Biochimica et biophysica acta (2014-07-27)
Juyeon Lee, Boyeon Park, Gayoung Kim, Kwangwoo Kim, Jeongjun Pak, Kwanhyeong Kim, Michael B Ye, Sung-Gyoo Park, Daeho Park
ZUSAMMENFASSUNG
Elmo is an evolutionarily conserved mammalian ortholog of Caenorhabditis elegans CED-12 with proposed roles during the removal of apoptotic cells, cell migration, neurite outgrowth, and myoblast fusion (Katoh and Negishi (2003) [1], Park and Tosello (2007) [2], Grimsley et al. (2004) [3], Hamoud et al. (2014) [4]). Elmo mediates these cellular processes by interacting with various proteins located in the plasma membrane, cytoplasm and nucleus, and by modulating their activities although it has no intrinsic catalytic activity (Park and Tosello (2007) [2], Hamoud et al. (2014) [4], Li et al. (2013) [5], Margaron, Fradet and Cote (2013) [6], and Mauldin et al. (2013)[7]). Because there are a limited number of proteins known to interact with Elmo, we performed a yeast two-hybrid screen using Elmo1 as bait to identify Elmo1-interacting proteins and to evaluate their mode of regulation. Arhgef16 was one of the proteins identified through the screen and subsequent analyses revealed that Arhgef16 interacted with Elmo1 in mammalian cells as well. Expression of Arhgef16 in phagocytes promoted engulfment of apoptotic cells, and engulfment mediated by Arhgef16 increased synergistically in the presence of Elmo1 but was abrogated in the absence of Elmo1. In addition, Arhgef16-mediated removal of apoptotic cells was dependent on RhoG, but independent of Dock1. Taken together, this study suggests that the newly identified Elmo1-interacting protein, Arhgef16, functions synergistically with Elmo1 to promote clearance of apoptotic cells in a RhoG-dependent and Dock1-independent manner.