Merck
  • Multicomponent linchpin couplings of silyl dithianes: synthesis of the Schreiber C(16-28) trisacetonide subtarget for mycoticins A and B.
Multicomponent linchpin couplings of silyl dithianes: synthesis of the Schreiber C(16-28) trisacetonide subtarget for mycoticins A and B.
Organic letters (2000-06-03)
A B Smith, S M Pitram
ZUSAMMENFASSUNG
[formula: see text] An efficient synthesis of trisacetonide (+)-11, the Schreiber C(16-28) subtarget for mycoticins A and B, is described. The key synthetic transformation entails a one-flask, five-component linchpin coupling tactic.