- Chromatographic profile of high boiling point organic acids in human urine.
Chromatographic profile of high boiling point organic acids in human urine.
Journal of chromatography (1978-03-01)
G K Brown, O Stokke, E Jellum
PMID649712
ZUSAMMENFASSUNG
The profile of high boiling point organic acids in urine samples from both normal subjects and patients suspected of having some form of metabolic disorder has been determined by combined gas chromatography-mass spectrometry. Fifteen different compounds eluting after hippuric acid have been identified, including two, cinnamoylglycine and acetyltributylcitrate, which have not been recognised previously. Relative retention times and abbreviated mass spectra of the identified compounds are presented.