Merck
  • Enantioselective hydrogenation of methyl acetoacetate catalyzed by nickel supported on activated carbon or graphite.
Enantioselective hydrogenation of methyl acetoacetate catalyzed by nickel supported on activated carbon or graphite.
Applied Catalysis A: General (2001)
Wolfson, A., et al.