Merck
Concise total synthesis of (+/-)-pseudotabersonine via double ring-closing metathesis strategy.
Organic letters (2010-08-14)
Bo Cheng, James D Sunderhaus, Stephen F Martin
ZUSAMMENFASSUNG
A concise synthesis of (+/-)-pseudotabersonine from commercially available 1-(phenylsulfonyl)-3-indolecarboxaldehyde has been accomplished. This synthesis features the convergent assembly of a key intermediate via a stepwise variant of a Mannich-type multicomponent coupling process, a double ring-closing metathesis, and a one-pot deprotection/cyclization reaction.