- Novel D-π-A structured Zn(II)-porphyrin dyes containing a bis(3,3-dimethylfluorenyl)amine moiety for dye-sensitised solar cells.
Novel D-π-A structured Zn(II)-porphyrin dyes containing a bis(3,3-dimethylfluorenyl)amine moiety for dye-sensitised solar cells.
Chemical communications (Cambridge, England) (2012-04-14)
Min Soo Kang, Sung Ho Kang, Sang Gyun Kim, In Taek Choi, Jung Ho Ryu, Myung Jong Ju, Daeheum Cho, Jin Yong Lee, Hwan Kyu Kim
PMID22499080
ZUSAMMENFASSUNG
A high solar-to-electricity conversion efficiency of 7.22% was achieved with a short circuit current (J(sc)) of 15.30 mA cm(-2), an open circuit voltage (V(oc)) of 669 mV and a fill factor (FF) of 0.71 for the 2Flu-ZnP-CN-COOH dye with a multi-functional co-adsorbent, under 100 mW cm(-2) AM 1.5 G simulated light.