- Spirally configured cis-stilbene/fluorene hybrids as bipolar, organic sensitizers for solar cell applications.
Spirally configured cis-stilbene/fluorene hybrids as bipolar, organic sensitizers for solar cell applications.
Chemical communications (Cambridge, England) (2012-04-14)
Wei-Shan Chao, Ken-Hsien Liao, Chien-Tien Chen, Wei-Kai Huang, Chi-Ming Lan, Eric Wei-Guang Diau
PMID22499126
ZUSAMMENFASSUNG
Hybrids based on a dibenzosuberene core bearing a spiro-fluorene junction at the C-5 position and with amino donor and β-thiophenyl-α-cyanoacrylic acid acceptor groups at C-3 and C-7, respectively, serve as new organic sensitizer materials for solar cell applications. Solar cell devices based on these materials show a conversion efficiency (η) of up to 6.1% (V(oc) = 697 mV, J(sc) = 12.2 mA cm(-2), FF = 0.72) under AM 1.5 G conditions. The best IPCE values exceed 75% within the 450-550 nm absorption range.