- A new class of histamine H(3)-receptor antagonists: synthesis and structure-activity relationships of 7,8,9,10-tetrahydro-6H-cyclohepta[b]quinolines.
A new class of histamine H(3)-receptor antagonists: synthesis and structure-activity relationships of 7,8,9,10-tetrahydro-6H-cyclohepta[b]quinolines.
Bioorganic & medicinal chemistry letters (2003-06-12)
Sean C Turner, Timothy A Esbenshade, Youssef L Bennani, Arthur A Hancock
PMID12798320
ZUSAMMENFASSUNG
The synthesis and biological evaluation of novel cycloheptaquinoline antagonists of the human H(3) receptor are described. Two series of compounds, bearing either an amino substituent or an alkyne linker at the 11-position, were investigated. Modifications of the amino substituents, optimization of chain length and the effect of conformational restraints are described. Several compounds with high affinity and selectivity for the H(3) receptor were discovered.