Merck
Synthesis of Antitumor Lycorines by Intramolecular Diels-Alder Reaction.
The Journal of organic chemistry (1996-03-08)
Dolores Pérez, Gema Burés, Enrique Guitián, Luis Castedo
ZUSAMMENFASSUNG
Pharmacologically interesting lycorines were obtained by a short, efficient method based on an intramolecular Diels-Alder reaction between an alpha-pyrone and an alkyne, followed by loss of CO(2) in a retro Diels-Alder reaction. The cyclization precursors (pyrones 9) were obtained in good yields in two or three steps from the corresponding homophthalic acid or anhydride.