Merck
  • Endothelial NOTCH1 is suppressed by circulating lipids and antagonizes inflammation during atherosclerosis.
Endothelial NOTCH1 is suppressed by circulating lipids and antagonizes inflammation during atherosclerosis.
The Journal of experimental medicine (2015-11-11)
Anaïs Briot, Mete Civelek, Atsuko Seki, Karen Hoi, Julia J Mack, Stephen D Lee, Jason Kim, Cynthia Hong, Jingjing Yu, Gregory A Fishbein, Ladan Vakili, Alan M Fogelman, Michael C Fishbein, Aldons J Lusis, Peter Tontonoz, Mohamad Navab, Judith A Berliner, M Luisa Iruela-Arispe
ZUSAMMENFASSUNG
Although much progress has been made in identifying the mechanisms that trigger endothelial activation and inflammatory cell recruitment during atherosclerosis, less is known about the intrinsic pathways that counteract these events. Here we identified NOTCH1 as an antagonist of endothelial cell (EC) activation. NOTCH1 was constitutively expressed by adult arterial endothelium, but levels were significantly reduced by high-fat diet. Furthermore, treatment of human aortic ECs (HAECs) with inflammatory lipids (oxidized 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphocholine [Ox-PAPC]) and proinflammatory cytokines (TNF and IL1β) decreased Notch1 expression and signaling in vitro through a mechanism that requires STAT3 activation. Reduction of NOTCH1 in HAECs by siRNA, in the absence of inflammatory lipids or cytokines, increased inflammatory molecules and binding of monocytes. Conversely, some of the effects mediated by Ox-PAPC were reversed by increased NOTCH1 signaling, suggesting a link between lipid-mediated inflammation and Notch1. Interestingly, reduction of NOTCH1 by Ox-PAPC in HAECs was associated with a genetic variant previously correlated to high-density lipoprotein in a human genome-wide association study. Finally, endothelial Notch1 heterozygous mice showed higher diet-induced atherosclerosis. Based on these findings, we propose that reduction of endothelial NOTCH1 is a predisposing factor in the onset of vascular inflammation and initiation of atherosclerosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ai848100