Merck
Ultratrace DNA Detection Based on the Condensing-Enrichment Effect of Superwettable Microchips.
Advanced materials (Deerfield Beach, Fla.) (2015-10-02)
Li-Ping Xu, Yanxia Chen, Gao Yang, Wanxin Shi, Bing Dai, Guannan Li, Yanhua Cao, Yongqiang Wen, Xueji Zhang, Shutao Wang
ZUSAMMENFASSUNG
A sensitive nucleic acid detection platform based on superhydrophilic microwells spotted on a superhydrophobic substrate is fabricated. Due to the wettability differences, ultratrace DNA molecules are enriched and the fluorescent signals are amplified to allow more sensitive detection. The biosensing interface based on superwettable materials provides a simple and cost-effective way for ultratrace DNA sensing.