Merck
Fer tyrosine kinase oligomer mediates and amplifies Src-induced tumor progression.
Oncogene (2015-04-14)
C Oneyama, Y Yoshikawa, Y Ninomiya, T Iino, S Tsukita, M Okada
ZUSAMMENFASSUNG
c-Src is upregulated in various human cancers, suggesting its role in malignant progression. However, the molecular circuits of c-Src oncogenic signaling remain elusive. Here we show that Fer tyrosine kinase oligomer mediates and amplifies Src-induced tumor progression. Previously, we showed that transformation of fibroblasts is promoted by the relocation of c-Src to non-raft membranes. In this study, we identified Fer and ezrin as non-raft c-Src targets. c-Src directly activated Fer by initiating its autophosphorylation, which was further amplified by Fer oligomerization. Fer interacted with active c-Src at focal adhesion membranes and activated Fer-phosphorylated ezrin to induce cell transformation. Fer was also crucial for cell transformation induced by v-Src or epidermal growth-factor receptor activation. Furthermore, Fer activation was required for tumorigenesis and invasiveness in some cancer cells in which c-Src is upregulated. We propose that the Src-Fer axis represents a new therapeutic target for treatment of a subset of human cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ezr