Merck
  • Phenolic content, antioxidant activity and effective compounds of kumquat extracted by different solvents.
Phenolic content, antioxidant activity and effective compounds of kumquat extracted by different solvents.
Food chemistry (2015-12-01)
Shyi-Neng Lou, Yi-Chun Lai, Ya-Siou Hsu, Chi-Tang Ho
ZUSAMMENFASSUNG
The total phenolic and flavonoid content of extracts from peel of kumquat were higher than those from pulp, and those extracted from immature kumquat were higher than those from mature kumquat. The highest levels of phenolic and flavonoid content were obtained in hot water extracts. The flavonoids of kumquat extracted from hot water were mainly soluble conjugated compounds, including C-glycosides, such as 3',5'-di-C-β-glucopyranosylphloretin (DGPP), acacetin 8-C-neohesperidoside (margaritene), acacetin 6-C-neohesperidoside (isomargaritene), apigenin 8-C-neohesperidoside, and O-glycosides, such as acacetin 7-O-neohesperidoside (fortunellin), isosakuranetin 7-O-neohesperidoside (poncirin) and apigenin 7-O-neohesperidoside (rhoifolin). A positive relationship existed between total phenolic content and DPPH scavenging potency (p<0.001). Total flavonoid content showed a similar correlation (p<0.001) to DPPH scavenging potency. The effective flavonoids contributing to antioxidant activity were DGPP and apigenin 8-C-neohesperidoside, which could be extracted in high amounts, by hot water at 90°C, from immature kumquat peel.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carbonsäure, 97%