Merck
Sodium calcium exchanger operates in the reverse mode in metastatic human melanoma cells.
Cellular and molecular biology (Noisy-le-Grand, France) (2015-11-17)
S R Sennoune, J M Santos, F Hussain, R Martínez-Zaguilán
ZUSAMMENFASSUNG
Cytosolic Ca2+ ([Ca2+]cyt) is important in the regulation of several cellular functions involved in metastasis. We hypothesize that distinct [Ca2+]cyt regulation explains the acquisition of a more metastatic phenotype. To test this hypothesis, we used highly and lowly metastatic human melanoma cells and [Ca2+]cyt was monitored using Fura—2AM and fluorescence spectroscopy. Stimulation with ATP elicited a sustained increase in [Ca2+]cyt in highly metastatic cells, but a transient increase in lowly metastatic cells. Na+ substitution revealed Na+/Ca2+ exchanger (NCX) activity in reverse mode in highly, but not in lowly metastatic cells. In highly metastatic cells, addition of Na+ in the plateau phase of [Ca2+]cyt increase elicited with ATP, in the absence of Na+, resulted in a rapid return to basal, indicating that NCX can operate in both reverse and forward modes. Inhibition and knockdown of NCX, using KB—R7943 and siRNA NCX—1 respectively, supported the significance of NCX in [Ca2+]cyt regulation in highly metastatic cells. Stimulation with UTP triggered a rapid increase in highly metastatic cells [Ca2+]cyt, but not in lowly metastatic cells suggesting that highly and lowly metastatic cells exhibit distinct purinergic receptors. These data indicate that following agonist—stimulation, NCX operates preferentially in the reverse mode to enable a sustained [Ca2+]cyt increase in highly metastatic cells. The forward mode of NCX operation to extrude Ca2+ is preferred in lowly metastatic cells. The acquisition of a more metastatic phenotype involves a switch in NCX activity from forward to reverse mode that is favorable to maintain elevated [Ca2+]cyt in response to agonist stimulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
KB-R7943, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tlx2