Merck
  • Inhomogeneity of collagen organization within the fibrotic scar after myocardial infarction: results in a swine model and in human samples.
Inhomogeneity of collagen organization within the fibrotic scar after myocardial infarction: results in a swine model and in human samples.
Journal of anatomy (2015-10-30)
Arantxa Hervas, Amparo Ruiz-Sauri, Elena de Dios, Maria Jose Forteza, Gema Minana, Julio Nunez, Cristina Gomez, Clara Bonanad, Nerea Perez-Sole, Jose Gavara, Francisco Javier Chorro, Vicente Bodi
ZUSAMMENFASSUNG
We aimed to characterize the organization of collagen within a fibrotic scar in swine and human samples from patients with chronic infarctions. Swine were subjected to occlusion of the left anterior descending artery followed by reperfusion 1 week (acute myocardial infarction group) or 1 month (chronic myocardial infarction group) after infarction. The organization of the collagen fibers (Fast Fourier Transform of samples after picrosirius staining; higher values indicate more disorganization) was studied in 100 swine and 95 human samples. No differences in collagen organization were found between the acute and chronic groups in the core area of the scar in the experimental model. In the chronic group, the endocardium [0.90 (0.84-0.94); median (interquartile range)], epicardium [0.84 (0.79-0.91)] and peripheral area [0.73 (0.63-0.83)] displayed a much more disorganized pattern than the core area of the fibrotic scar [0.56 (0.45-0.64)]. Similarly, in human samples, the collagen fibers were more disorganized in all of the outer areas than in the core of the fibrotic scar (P < 0.0001). Both in a highly controlled experimental model and in patient samples, collagen fibers exhibited an organized pattern in the core of the infarction, whereas the outer areas displayed a high level of inhomogeneity. This finding contributes pathophysiological information regarding the healing process and may lead to a clearer understanding of the genesis and invasive treatment of arrhythmias after acute myocardial infarction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
2,3,5-Triphenyl-tetrazoliumchlorid -Lösung, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC