Merck
  • Molecular cloning and characterization of Siamese crocodile (Crocodylus siamensis) copper, zinc superoxide dismutase (CSI-Cu,Zn-SOD) gene.
Molecular cloning and characterization of Siamese crocodile (Crocodylus siamensis) copper, zinc superoxide dismutase (CSI-Cu,Zn-SOD) gene.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology (2015-11-03)
Penporn Sujiwattanarat, Parinya Pongsanarakul, Yosapong Temsiripong, Theeranan Temsiripong, Charin Thawornkuno, Yoshinobu Uno, Sasimanas Unajak, Yoichi Matsuda, Kiattawee Choowongkomon, Kornsorn Srikulnath
ZUSAMMENFASSUNG
Superoxide dismutase (SOD, EC 1.15.1.1) is an antioxidant enzyme found in all living cells. It regulates oxidative stress by breaking down superoxide radicals to oxygen and hydrogen peroxide. A gene coding for Cu,Zn-SOD was cloned and characterized from Siamese crocodile (Crocodylus siamensis; CSI). The full-length expressed sequence tag (EST) of this Cu,Zn-SOD gene (designated as CSI-Cu,Zn-SOD) contained 462bp encoding a protein of 154 amino acids without signal peptides, indicated as intracellular CSI-Cu,Zn-SOD. This agreed with the results from the phylogenetic tree, which indicated that CSI-Cu,Zn-SOD belonged to the intracellular Cu,Zn-SOD. Chromosomal location determined that the CSI-Cu,Zn-SOD was localized to the proximal region of the Siamese crocodile chromosome 1p. Several highly conserved motifs, two conserved signature sequences (GFHVHEFGDNT and GNAGGRLACGVI), and conserved amino acid residues for binding copper and zinc (His(47), His(49), His(64), His(72), His(81), Asp(84), and His(120)) were also identified in CSI-Cu,Zn-SOD. Real-time PCR analysis showed that CSI-Cu,Zn-SOD mRNA was expressed in all the tissues examined (liver, pancreas, lung, kidney, heart, and whole blood), which suggests a constitutively expressed gene in these tissues. Expression of the gene in Escherichia coli cells followed by purification yielded a recombinant CSI-Cu,Zn-SOD, with Km and Vmax values of 6.075mM xanthine and 1.4×10(-3)mmolmin(-1)mg(-1), respectively. This Vmax value was 40 times lower than native Cu,Zn-SOD (56×10(-3)mmolmin(-1)mg(-1)), extracted from crocodile erythrocytes. This suggests that cofactors, protein folding properties, or post-translational modifications were lost during the protein purification process, leading to a reduction in the rate of enzyme activity in bacterial expression of CSI-Cu,Zn-SOD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
E-64d, protease inhibitor