- Characteristics of silver nanoparticles in vehicles for biological applications.
Characteristics of silver nanoparticles in vehicles for biological applications.
International journal of pharmaceutics (2015-10-13)
Kristina Kejlová, Věra Kašpárková, Daniel Krsek, Dagmar Jírová, Hana Kolářová, Markéta Dvořáková, Kateřina Tománková, Veronika Mikulcová
PMID26456248
ZUSAMMENFASSUNG
Silver nanoparticles (AgNPs) have been used for decades as anti-bacterial agents in various industrial fields such as cosmetics, health industry, food storage, textile coatings and environmental applications, although their toxicity is not fully recognized yet. Antimicrobial and catalytic activity of AgNPs depends on their size as well as structure, shape, size distribution, and physico-chemical environment. The unique properties of AgNPs require novel or modified toxicological methods for evaluation of their toxic potential combined with robust analytical methods for characterization of nanoparticles applied in relevant vehicles, e.g., culture medium with/without serum and phosphate buffered saline.
MATERIALIEN