Merck
  • Diet-Induced Obesity Enhances Progression of Hepatocellular Carcinoma through Tenascin-C/Toll-Like Receptor 4 Signaling.
Diet-Induced Obesity Enhances Progression of Hepatocellular Carcinoma through Tenascin-C/Toll-Like Receptor 4 Signaling.
The American journal of pathology (2015-11-26)
Jennifer H Benbow, Kyle J Thompson, Heidi L Cope, Elizabeth Brandon-Warner, Catherine R Culberson, Krista L Bossi, Ting Li, Mark W Russo, Keith S Gersin, Iain H McKillop, Andrew S deLemos, Laura W Schrum
ZUSAMMENFASSUNG
Obesity is an independent risk factor for the development of liver fibrosis/cirrhosis and hepatocellular carcinoma (HCC). Tenascin-C (TnC), an extracellular matrix protein, is transiently expressed during tissue injury and plays a role in fibrogenesis and tumorigenesis. However, the mechanistic role of TnC signaling in the development of HCC remains unknown. We developed a diet-induced obesity HCC mouse model and examined TnC expression and liver injury. To determine the cellular mechanism of TnC signaling in promoting inflammation and hepatocyte epithelial-mesenchymal transition and migration, we used primary hepatocytes and hepatoma and macrophage cell lines. Further, to determine whether elevated TnC expression correlated with obesity-associated HCC, we measured plasma TnC in obese patients with various levels of liver injury. Increased tissue inflammation accompanied with elevated hepatic stellate cell-derived TnC and Toll-like receptor 4 expression was observed in the diet-induced obesity HCC animal model. In vitro studies found enhanced Toll-like receptor 4 signaling activated by TnC, promoting an increased inflammatory response, hepatocyte transformation, and migration. Further, obese patients with cirrhosis alone and in combination with HCC showed significant increases in plasma TnC compared with healthy volunteers and patients with less severe liver injury. Overall, these studies suggest TnC/Toll-like receptor 4 signaling as an important regulator in HCC; inhibiting this signaling axis may be a viable therapeutic target for impeding HCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O