Merck
  • Genetic variants in the renin-angiotensin system predict response to bevacizumab in cancer patients.
Genetic variants in the renin-angiotensin system predict response to bevacizumab in cancer patients.
European journal of clinical investigation (2015-10-29)
Diana Moreno-Muñoz, Juan R de la Haba-Rodríguez, Francisco Conde, Laura M López-Sánchez, Araceli Valverde, Vanessa Hernández, Antonio Martínez, Carlos Villar, Auxiliadora Gómez-España, Ignacio Porras, Antonio Rodríguez-Ariza, Enrique Aranda
ZUSAMMENFASSUNG
Currently, there are no predictive biomarkers for anti-angiogenic strategies in cancer, but response to anti-angiogenic drugs is associated with development of hypertension secondary to treatment. Therefore, this study explored the clinical relevance of genetic polymorphisms in some components of the renin-angiotensin system (RAS). Genomic DNA was isolated from peripheral blood from 95 metastatic breast or colorectal cancer patients treated with bevacizumab, and AGTR1-A1166C (rs5186), AGT-M235T (rs699) SNPs and ACE I/D (rs4646994) polymorphisms were genotyped using RT-PCR. Circulating vascular endothelial grow factor and angiotensin converting enzyme (ACE) levels were analysed using ELISA kits. The antitumoral activity of bevacizumab was assayed in mice orthotopically xenografted with AGTR1-overexpressing breast cancer cells. The ACE IN/IN genotype was associated with a higher rate of disease progression compared to DEL/IN and DEL/DEL genotypes (36% vs. 11·1% P < 0·05). Similarly, AGTR1-1166A/A genotype was also associated with a higher rate of disease progression compared to AGTR1-1166A/C and AGTR1-1166C/C genotypes (24·4% vs. 2·7% P < 0·01). ACE IN/IN genotype was also found to be associated with shorter time to treatment failure compared to ACE IN/DEL and ACE DEL/DEL genotypes (14 weeks vs. 41·71, P = 0·033), whereas circulating ACE levels were found to be associated with a better response to bevacizumab treatment. Besides, in vivo experiments showed a significantly higher antitumoral activity of bevacizumab in tumours derived from AGTR1-overexpressing breast cancer cells. A higher activity of ACE-angiotensin-II-AGTR1 axis is associated with a better response to bevacizumab, supporting that the RAS can be an important source of potential predictive markers of response to anti-angiogenic drugs.