Merck
  • An Open-Label, Randomized Phase II Trial of Personalized Peptide Vaccination in Patients with Bladder Cancer that Progressed after Platinum-Based Chemotherapy.
An Open-Label, Randomized Phase II Trial of Personalized Peptide Vaccination in Patients with Bladder Cancer that Progressed after Platinum-Based Chemotherapy.
Clinical cancer research : an official journal of the American Association for Cancer Research (2015-11-20)
Masanori Noguchi, Kazumasa Matsumoto, Hirotsugu Uemura, Gaku Arai, Masatoshi Eto, Seiji Naito, Chikara Ohyama, Yasutomo Nasu, Masatoshi Tanaka, Fukuko Moriya, Shigetaka Suekane, Satoko Matsueda, Nobukazu Komatsu, Tetsuro Sasada, Akira Yamada, Tatsuyuki Kakuma, Kyogo Itoh
ZUSAMMENFASSUNG
The prognosis of platinum-based chemotherapy-resistant metastatic urothelial cancer of the bladder remains poor. Personalized selection of the right peptides for each patient could be a novel approach for a cancer vaccine to boost anticancer immunity. In this randomized, open-label, phase II study, patients ages ≥18 years with progressive bladder cancer after first-line platinum-based chemotherapy were randomly assigned (1:1) to receive personalized peptide vaccination (PPV) plus best supportive care (BSC) or BSC. PPV treatment used a maximum of four peptides chosen from 31 candidate peptides according to human leukocyte antigen types and peptide-reactive IgG titers, for 12 s.c. injections (8 injections, weekly; 4 injections, bi-weekly). The primary endpoint was progression-free survival (PFS). Secondary endpoints were overall survival (OS), immune response, and toxicity. Eighty patients were randomly assigned to receive either PPV plus BSC (n = 39) or BSC (n = 41). No significant improvement in PFS was noted [HR, 0.7; 95% confidence interval (CI), 0.4-1.2, P = 0.17]. For the secondary endpoints, PPV plus BSC significantly prolonged OS compared with BSC (HR, 0.58; 95% CI, 0.34-0.99, P = 0.049), with median OS of 7.9 months (95% CI, 3.5-12.0) in the PPV plus BSC and 4.1 months (95% CI, 2.8-6.9) in the BSC. PPV treatment was well tolerated, without serious adverse drug reactions. PPV could not prolong PFS, but OS appeared to be improved with low toxicity and immune responses. Further large-scale, randomized trials are needed to confirm these results.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,O-Bis-(trimethylsilyl)-carbamat, ≥98.0% (T)