Merck
  • Modeling the Dynamic AMD-Associated Chronic Oxidative Stress Changes in Human ESC and iPSC-Derived RPE Cells.
Modeling the Dynamic AMD-Associated Chronic Oxidative Stress Changes in Human ESC and iPSC-Derived RPE Cells.
Investigative ophthalmology & visual science (2015-11-26)
Thelma Y Garcia, Mark Gutierrez, Joseph Reynolds, Deepak A Lamba
ZUSAMMENFASSUNG
Here we use human embryonic stem cells (hESCs) and human-induced pluripotent stem cell (hiPSC)-derived retinal pigment epithelium (RPE) cells to model chronic oxidative stress in vitro. This model allows us to understand the evolution of chronic stress response in RPE in vivo, as well as to monitor microRNAs changes. Finally, we use this in vitro model to identify a partial agonist of NRF2 that is protective against reactive oxygen species (ROS)-induced cytotoxicity. The hESCs and hiPSCs were differentiated toward an RPE fate. Upon maturation, RPE cells were subjected to chronic oxidative stress using Paraquat (PQ). The cells were then analyzed using immunocytochemistry and quantitative RT-PCR to look for changes in gene expression and microRNA changes. Small molecules targeting NRF2 pathways were utilized to look for protection against oxidative stress-induced apoptosis. We show that 160 μM PQ can be used to generate a model of chronic oxidative stress in RPE cells derived from hESCs and hiPSCs. Using this model, we characterize the NRF2 pathway effectors during the early and late stages of chronic oxidative stress and identify microRNAs changes during oxidative stress. We find that hsa-miR144 modulates NRF2 activity during ROS stress. Lastly, we found a small molecule modulator of NRF2 that plays a protective role against oxidative stress-induced RPE apoptosis. In summary, pluripotent stem cell-derived retinal cells can be used to model retinal diseases in a dish. This can provide an unprecedented opportunity to understand the evolution of disease processes and allow us to identify novel therapeutics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
2-Phenylindol, technical grade, 95%