Merck
  • The erbB3- and IGF-1 receptor-initiated signaling pathways exhibit distinct effects on lapatinib sensitivity against trastuzumab-resistant breast cancer cells.
The erbB3- and IGF-1 receptor-initiated signaling pathways exhibit distinct effects on lapatinib sensitivity against trastuzumab-resistant breast cancer cells.
Oncotarget (2015-12-02)
Hui Lyu, Xiao He Yang, Susan M Edgerton, Ann D Thor, Xiaoying Wu, Zhimin He, Bolin Liu
ZUSAMMENFASSUNG
Both erbB3 and IGF-1 receptor (IGF-1R) have been shown to play an important role in trastuzumab resistance. However, it remains unclear whether erbB3- and IGF-1R-initiated signaling pathways possess distinct effects on the sensitivity of lapatinib, a dual tyrosine kinase inhibitor against both EGFR and erbB2, in trastuzumab-resistant breast cancer. Here, we show that the trastuzumab-resistant SKBR3-pool2 and BT474-HR20 breast cancer sublines, as compared the parental SKBR3 and BT474 cells, respectively, exhibit refractoriness to lapatinib. Knockdown of erbB3 inhibited Akt in SKBR3-pool2 and BT474-HR20 cells, significantly increased lapatinib efficacy, and dramatically re-sensitized the cells to lapatinib-induced apoptosis. In contrast, specific knockdown of IGF-1R did not alter the cells' responsiveness to lapatinib. While the levels of phosphorylated Src (P-Src) were reduced upon IGF-1R downregulation, the P-Akt levels remained unchanged. Furthermore, a specific inhibitor of Akt, but not Src, significantly enhanced lapatinib-mediated anti-proliferative/anti-survival effects on SKBR3-pool2 and BT474-HR20 cells. These data indicate that erbB3 signaling is critical for both trastuzumab and lapatinib resistances mainly through the PI-3K/Akt pathway, whereas IGF-1R-initiated Src activation results in trastuzumab resistance without affecting lapatinib sensitivity. Our findings may facilitate the development of precision therapeutic regimens for erbB2-positive breast cancer patients who become resistant to erbB2-targeted therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
Akt1/2 kinase inhibitor, ≥98% (HPLC)