Merck
  • Motor development related to duration of exclusive breastfeeding, B vitamin status and B12 supplementation in infants with a birth weight between 2000-3000 g, results from a randomized intervention trial.
Motor development related to duration of exclusive breastfeeding, B vitamin status and B12 supplementation in infants with a birth weight between 2000-3000 g, results from a randomized intervention trial.
BMC pediatrics (2015-12-19)
Ingrid Kristin Torsvik, Per Magne Ueland, Trond Markestad, Øivind Midttun, Anne-Lise Bjørke Monsen
ZUSAMMENFASSUNG
Exclusive breastfeeding for 6 months is assumed to ensure adequate micronutrients for term infants. Our objective was to investigate the effects of prolonged breastfeeding on B vitamin status and neurodevelopment in 80 infants with subnormal birth weights (2000-3000 g) and examine if cobalamin supplementation may benefit motor function in infants who developed biochemical signs of impaired cobalamin function (total homocysteine (tHcy) > 6.5 μmol/L) at 6 months. Levels of cobalamin, folate, riboflavin and pyridoxal 5´-phosphate, and the metabolic markers tHcy and methylmalonic acid (MMA), were determined at 6 weeks, 4 and 6 months (n = 80/68/66). Neurodevelopment was assessed with the Alberta Infants Motor Scale (AIMS) and the parental questionnaire Ages and Stages (ASQ) at 6 months. At 6 months, 32 of 36 infants with tHcy > 6.5 μmol/L were enrolled in a double blind randomized controlled trial to receive 400 μg hydroxycobalamin intramuscularly (n = 16) or sham injection (n = 16). Biochemical status and neurodevelopment were evaluated after one month. Except for folate, infants who were exclusively breastfed for >1 month had lower B vitamin levels at all assessments and higher tHcy and MMA levels at 4 and 6 months. At 6 months, these infants had lower AIMS scores (p = 0.03) and ASQ gross motor scores (p = 0.01). Compared to the placebo group, cobalamin treatment resulted in a decrease in plasma tHcy (p < 0.001) and MMA (p = 0.001) levels and a larger increase in AIMS (p = 0.02) and ASQ gross motor scores (p = 0.03). The findings suggest that prolonged exclusive breastfeeding may not provide sufficient B vitamins for small infants, and that this may have a negative effect on early gross motor development. In infants with mild cobalamin deficiency at 6 months, cobalamin treatment significantly improvement cobalamin status and motor function, suggesting that the observed impairment in motor function associated with long-term exclusive breastfeeding, may be due to cobalamin deficiency. ClinicalTrials.gov, number NCT01201005.