Merck
  • CD14 as a Mediator of the Mineralocorticoid Receptor-Dependent Anti-apolipoprotein A-1 IgG Chronotropic Effect on Cardiomyocytes.
CD14 as a Mediator of the Mineralocorticoid Receptor-Dependent Anti-apolipoprotein A-1 IgG Chronotropic Effect on Cardiomyocytes.
Endocrinology (2015-09-24)
Tiphaine Mannic, Nathalie Satta, Sabrina Pagano, Magaly Python, Julien Virzi, Fabrizio Montecucco, Miguel A Frias, Richard W James, Andres D Maturana, Michel F Rossier, Nicolas Vuilleumier
ZUSAMMENFASSUNG
In vitro and animal studies point to autoantibodies against apolipoprotein A-1 (anti-apoA-1 IgG) as possible mediators of cardiovascular (CV) disease involving several mechanisms such as basal heart rate interference mediated by a mineralocorticoid receptor-dependent L-type calcium channel activation, and a direct pro-inflammatory effect through the engagement of the toll-like receptor (TLR) 2/CD14 complex. Nevertheless, the possible implication of these receptors in the pro-arrhythmogenic effect of anti-apoA-1 antibodies remains elusive. We aimed at determining whether CD14 and TLRs could mediate the anti-apoA-1 IgG chronotropic response in neonatal rat ventricular cardiomyocytes (NRVC). Blocking CD14 suppressed anti-apoA-1 IgG binding to NRVC and the related positive chronotropic response. Anti-apoA-1 IgG alone induced the formation of a TLR2/TLR4/CD14 complex, followed by the phosphorylation of Src, whereas aldosterone alone promoted the phosphorylation of Akt by phosphatidylinositol 3-kinase (PI3K), without affecting the chronotropic response. In the presence of both aldosterone and anti-apoA-1 IgG, the localization of TLR2/TLR4/CD14 was increased in membrane lipid rafts, followed by PI3K and Src activation, leading to an L-type calcium channel-dependent positive chronotropic response. Pharmacological inhibition of the Src pathway led to the decrease of L-type calcium channel activity and abrogated the NRVC chronotropic response. Activation of CD14 seems to be a key regulator of the mineralocorticoid receptor-dependent anti-apoA-1 IgG positive chronotropic effect on NRVCs, involving relocation of the CD14/TLR2/TLR4 complex into lipid rafts followed by PI3K and Src-dependent L-type calcium channel activation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Laminin aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dicyanocobinamid, ≥93%
Sigma-Aldrich
PP2, ≥98% (HPLC)