Merck
Signal Transduction and Intracellular Trafficking by the Interleukin 36 Receptor.
The Journal of biological chemistry (2015-08-14)
Siddhartha S Saha, Divyendu Singh, Ernest L Raymond, Rajkumar Ganesan, Gary Caviness, Christine Grimaldi, Joseph R Woska, Detlev Mennerich, Su-Ellen Brown, M Lamine Mbow, C Cheng Kao
ZUSAMMENFASSUNG
Improper signaling of the IL-36 receptor (IL-36R), a member of the IL-1 receptor family, has been associated with various inflammation-associated diseases. However, the requirements for IL-36R signal transduction remain poorly characterized. This work seeks to define the requirements for IL-36R signaling and intracellular trafficking. In the absence of cognate agonists, IL-36R was endocytosed and recycled to the plasma membrane. In the presence of IL-36, IL-36R increased accumulation in LAMP1+ lysosomes. Endocytosis predominantly used a clathrin-mediated pathway, and the accumulation of the IL-36R in lysosomes did not result in increased receptor turnover. The ubiquitin-binding Tollip protein contributed to IL-36R signaling and increased the accumulation of both subunits of the IL-36R.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by ion-exchange chromatography, TLR ligand tested