Merck
  • Arsenic trioxide stimulates SUMO-2/3 modification leading to RNF4-dependent proteolytic targeting of PML.
Arsenic trioxide stimulates SUMO-2/3 modification leading to RNF4-dependent proteolytic targeting of PML.
FEBS letters (2008-08-19)
Stefan R Weisshaar, Kirstin Keusekotten, Anke Krause, Christiane Horst, Helen M Springer, Kerstin Göttsche, R Jürgen Dohmen, Gerrit J K Praefcke
ZUSAMMENFASSUNG
We have recently reported that poly-SUMO-2/3 conjugates are subject to a ubiquitin-dependent proteolytic control in human cells. Here we show that arsenic trioxide (ATO) increases SUMO-2/3 modification of promyelocytic leukemia (PML) leading to its subsequent ubiquitylation in vivo. The SUMO-binding ubiquitin ligase RNF4 mediates this modification and causes disruption of PML nuclear bodies upon treatment with ATO. Reconstitution of SUMO-dependent ubiquitylation of PML by RNF4 in vitro and in a yeast trans vivo system revealed a preference of RNF4 for chain forming SUMOs. Polysumoylation of PML in response to ATO thus leads to its recognition and ubiquitylation by RNF4.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution