Merck
  • Four bridging bis chelate ligands with very low lying. Π* orbitals. MO perturbation calculations, electrochemistry, and spectroscopy of mononuclear and binuclear group 6 metal tetracarbonyl complexes.
Four bridging bis chelate ligands with very low lying. Π* orbitals. MO perturbation calculations, electrochemistry, and spectroscopy of mononuclear and binuclear group 6 metal tetracarbonyl complexes.
Inorganic Chemistry (1987)
Kaim W and Kohlmann S.