Merck
  • The Impact of LEP G-2548A and LEPR Gln223Arg Polymorphisms on Adiposity, Leptin, and Leptin-Receptor Serum Levels in a Mexican Mestizo Population.
The Impact of LEP G-2548A and LEPR Gln223Arg Polymorphisms on Adiposity, Leptin, and Leptin-Receptor Serum Levels in a Mexican Mestizo Population.
BioMed research international (2015-06-13)
Efraín Chavarria-Avila, Mónica Vázquez-Del Mercado, Eduardo Gomez-Bañuelos, Sandra-Luz Ruiz-Quezada, Jorge Castro-Albarran, Lizeth Sánchez-López, Beatriz Teresita Martín-Marquez, Rosa-Elena Navarro-Hernández
ZUSAMMENFASSUNG
The polymorphisms in leptin (LEP G-2548A) and leptin-receptor (LEPR Gln223Arg) seem to influence obesity and lipid metabolism among others. The aim of this study was to investigate the effect of these polymorphisms on adiposity, leptin (sLeptin), and leptin-receptor (sLeptin-receptor) serum concentrations as well as inflammation markers. We included 382 adults originally from Western Mexico. They were genotyped by PCR-RFLP. Obese individuals showed higher sLeptin (58.2 ± 31.35 ng/mL) but lower sLeptin-receptor (12.6 ± 3.74 ng/mL) levels than normal weight ones (17.6 ± 14.62 ng/mL, 17.4 ± 4.62 ng/mL, resp.), P < 0.001. Obese subjects carriers of Arg/Arg genotype had more (P = 0.016) sLeptin-receptor (14.7 ± 4.96 ng/mL) and less (P = 0.004) sLeptin (44.0 ± 28.12 ng/mL) levels than Gln/Gln genotype (11.0 ± 2.92 ng/mL, 80.3 ± 33.24 ng/mL, resp.). Body fat mass was lower (P from 0.003 to 0.045) for A/A (36.5% ± 6.80) or Arg/Arg (36.8% ± 6.82) genotypes with respect to G/G (41.3% ± 5.52) and G/A (41.6% ± 5.61) or Gln/Gln (43.7% ± 4.74) and Gln/Arg (41.0% ± 5.52) genotypes carriers. Our results suggest that LEP -2548A and LEPR 223Arg could be genetic markers of less body fat mass accumulation in obese subjects from Western Mexico.